Assay Details
| Gene Symbol: Immt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844581.3 | XP_004844638.1 | 8 - 9 | 1005 | 59 | |
| XM_004844582.3 | XP_004844639.1 | 8 - 9 | 933 | 59 | ||
| XM_004844584.3 | XP_004844641.1 | 7 - 8 | 943 | 59 | ||
| XM_004844587.3 | XP_004844644.1 | 6 - 7 | 734 | 59 | ||
| XM_021258204.1 | XP_021113863.1 | 8 - 9 | 932 | 59 | ||
| XM_021258205.1 | XP_021113864.1 | 7 - 8 | 838 | 59 | ||
More Information
Set Membership: |
|

Immt
Genomic Map