Assay Details
| Gene Symbol: Ston1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839419.2 | XP_004839476.1 | 3 - 4 | 2223 | 69 | |
| XM_013070223.2 | XP_012925677.1 | 3 - 4 | 2398 | 69 | ||
| XM_013070224.2 | XP_012925678.1 | 3 - 4 | 2429 | 69 | ||
| XM_013070225.2 | XP_012925679.1 | 3 - 4 | 2439 | 69 | ||
More Information
Set Membership: |
|

Ston1
Genomic Map