Assay Details
| Gene Symbol: Cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845498.3 | XP_004845555.1 | 4 - 5 | 420 | 68 | |
| XM_004845499.3 | XP_004845556.1 | 4 - 5 | 389 | 68 | ||
| XM_021258913.1 | XP_021114572.1 | 4 - 5 | 496 | 68 | ||
More Information
Set Membership: |
|

Cenpm
Genomic Map