Assay Details
| Gene Symbol: Dynlrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842810.2 | XP_004842867.1 | 3 - 4 | 387 | 80 | |
| XM_021256678.1 | XP_021112337.1 | 3 - 4 | 396 | 80 | ||
More Information
Set Membership: |
|

Dynlrb2
Genomic Map