Assay Details
| Gene Symbol: Ocrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021240573.1 | XP_021096232.1 | 13 - 14 | 1555 | 59 | |
| XM_021240574.1 | XP_021096233.1 | 13 - 14 | 1555 | 59 | ||
| XM_021240575.1 | XP_021096234.1 | 13 - 14 | 1555 | 59 | ||
More Information
Set Membership: |
|

Ocrl
Genomic Map