Assay Details
Gene Symbol: Piga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866087.2 | XP_004866144.1 | 3 - 4 | 2173 | 65 | |
XM_004866088.2 | XP_004866145.1 | 3 - 4 | 1510 | 65 | ||
XM_004866089.2 | XP_004866146.1 | 3 - 4 | 2176 | 65 | ||
XM_004866090.2 | XP_004866147.1 | 3 - 4 | 1477 | 65 | ||
XM_021245358.1 | XP_021101017.1 | 3 - 4 | 2182 | 65 | ||
XM_021245359.1 | XP_021101018.1 | 2 - 3 | 630 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
