Assay Details
| Gene Symbol: Klf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850239.3 | XP_004850296.1 | 2 - 3 | 383 | 106 | |
| XM_021262744.1 | XP_021118403.1 | 3 - 4 | 630 | 106 | ||
More Information
Set Membership: |
|

Klf10
Genomic Map