Assay Details
| Gene Symbol: Retreg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859319.2 | XP_004859376.2 | 6 - 7 | 822 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Retreg3| Gene Symbol: Retreg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859319.2 | XP_004859376.2 | 6 - 7 | 822 | 75 | |