Assay Details
| Gene Symbol: LOC101720535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845834.3 | XP_004845891.1 | 1 | 121 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004845834.3 transcripts.
Set Membership: |
|

LOC101720535
Genomic Map