Assay Details
| Gene Symbol: Gemin6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839327.2 | XP_004839384.1 | 1 - 2 | 163 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004839327.2 transcripts.
Set Membership: |
|

Gemin6
Genomic Map