Assay Details
| Gene Symbol: Gemin6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839327.2 | XP_004839384.1 | 2 - 3 | 300 | 96 | |
| XM_004839328.2 | XP_004839385.1 | 2 - 3 | 247 | 96 | ||
| XM_021253741.1 | XP_021109400.1 | 1 - 2 | 1136 | 96 | ||
More Information
Set Membership: |
|

Gemin6
Genomic Map