Assay Details
| Gene Symbol: Dusp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867413.2 | XP_004867470.1 | 3 - 4 | 2317 | 67 | |
| XM_021246431.1 | XP_021102090.1 | 4 - 5 | 2470 | 67 | ||
More Information
Set Membership: |
|

Dusp10
Genomic Map