Assay Details
| Gene Symbol: Esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844195.2 | XP_004844252.1 | 5 - 6 | 450 | 72 | |
| XM_004844196.2 | XP_004844253.1 | 5 - 6 | 452 | 72 | ||
| XM_004844197.2 | XP_004844254.1 | 4 - 5 | 351 | 72 | ||
More Information
Set Membership: |
|

Esd
Genomic Map