Assay Details
| Gene Symbol: Fosl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839228.2 | XP_004839285.1 | 3 - 4 | 665 | 73 | |
| XM_004839229.3 | XP_004839286.1 | 3 - 4 | 4784 | 73 | ||
| XM_004839230.3 | XP_004839287.1 | 3 - 4 | 985 | 73 | ||
| XM_021253644.1 | XP_021109303.1 | 4 - 5 | 911 | 73 | ||
More Information
Set Membership: |
|

Fosl2
Genomic Map