Assay Details
| Gene Symbol: G6pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859293.3 | XP_004859350.1 | 4 - 5 | 635 | 68 | |
| XM_004859295.3 | XP_004859352.1 | 4 - 5 | 449 | 68 | ||
| XM_004859296.3 | XP_004859353.1 | 4 - 5 | 465 | 68 | ||
More Information
Set Membership: |
|

G6pc
Genomic Map