Assay Details
| Gene Symbol: LOC101700661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871024.3 | XP_004871081.1 | 4 - 5 | 672 | 116 | |
| XM_004871025.3 | XP_004871082.1 | 4 - 5 | 624 | 116 | ||
| XM_004871026.3 | XP_004871083.1 | 4 - 5 | 642 | 116 | ||
| XM_004871027.3 | XP_004871084.1 | 4 - 5 | 623 | 116 | ||
| XM_004871028.3 | XP_004871085.1 | 4 - 5 | 526 | 116 | ||
More Information
Set Membership: |
|

LOC101700661
Genomic Map