Assay Details
| Gene Symbol: Gtf2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835636.2 | XP_004835693.1 | 4 - 5 | 1075 | 91 | |
| XM_004835637.2 | XP_004835694.1 | 4 - 5 | 1081 | 91 | ||
More Information
Set Membership: |
|

Gtf2e1
Genomic Map