Assay Details
Gene Symbol: Tmem170a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842835.3 | XP_004842892.2 | 1 - 2 | 409 | 64 | |
XM_004842836.3 | XP_004842893.2 | 1 - 2 | 406 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
