Assay Details
| Gene Symbol: Znhit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870395.3 | XP_004870452.1 | 3 - 4 | 1286 | 91 | |
| XM_004870397.3 | XP_004870454.1 | 5 - 6 | 564 | 91 | ||
| XM_021248670.1 | XP_021104329.1 | 4 - 5 | 559 | 91 | ||
More Information
Set Membership: |
|

Znhit3
Genomic Map