Assay Details
| Gene Symbol: Wee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263712.1 | XP_021119371.1 | 1 - 2 | 856 | 58 | |
| XM_021263713.1 | XP_021119372.1 | 1 - 2 | 855 | 58 | ||
| XM_021263714.1 | XP_021119373.1 | 1 - 2 | 851 | 58 | ||
| XM_021263715.1 | XP_021119374.1 | 2 - 3 | 544 | 58 | ||
More Information
Set Membership: |
|

Wee2
Genomic Map