Assay Details
| Gene Symbol: Pimreg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857264.2 | XP_004857321.1 | 2 - 3 | 713 | 78 | |
| XM_013076762.2 | XP_012932216.1 | 2 - 3 | 714 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pimreg| Gene Symbol: Pimreg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857264.2 | XP_004857321.1 | 2 - 3 | 713 | 78 | |
| XM_013076762.2 | XP_012932216.1 | 2 - 3 | 714 | 78 | ||