Assay Details
| Gene Symbol: LOC101706826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065184.2 | XP_012920638.1 | 5 - 6 | 472 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101706826| Gene Symbol: LOC101706826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065184.2 | XP_012920638.1 | 5 - 6 | 472 | 81 | |
Set Membership: |
|