Assay Details
| Gene Symbol: Bud31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840516.3 | XP_004840573.1 | 3 - 4 | 645 | 81 | |
| XM_004840517.2 | XP_004840574.1 | 3 - 4 | 454 | 81 | ||
| XM_004840519.3 | XP_004840576.1 | 3 - 4 | 439 | 81 | ||
| XM_013070654.2 | XP_012926108.1 | 3 - 4 | 408 | 81 | ||
| XM_013070655.2 | XP_012926109.1 | 3 - 4 | 390 | 81 | ||
| XM_021254843.1 | XP_021110502.1 | 4 - 5 | 546 | 81 | ||
| XM_021254844.1 | XP_021110503.1 | 2 - 3 | 192 | 81 | ||
More Information
Set Membership: |
|

Bud31
Genomic Map