Assay Details
| Gene Symbol: Dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021250327.1 | XP_021105986.1 | 2 - 3 | 910 | 106 | |
| XM_021250329.1 | XP_021105988.1 | 3 - 4 | 936 | 106 | ||
More Information
Set Membership: |
|

Dnajb1
Genomic Map