Assay Details
| Gene Symbol: Mthfs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874809.3 | XP_004874866.1 | 1 - 2 | 182 | 83 | |
| XM_021252188.1 | XP_021107847.1 | 1 - 2 | 196 | 83 | ||
More Information
Set Membership: |
|

Mthfs
Genomic Map