Assay Details
| Gene Symbol: Fsd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848184.3 | XP_004848241.1 | 12 - 13 | 1558 | 79 | |
| XM_013073479.2 | XP_012928933.1 | 13 - 14 | 1591 | 79 | ||
| XM_013073480.2 | XP_012928934.1 | 12 - 13 | 1555 | 79 | ||
| XM_013073481.2 | XP_012928935.1 | 12 - 13 | 1367 | 79 | ||
| XM_013073482.2 | XP_012928936.1 | 11 - 12 | 1363 | 79 | ||
More Information
Set Membership: |
|

Fsd1l
Genomic Map