Assay Details
| Gene Symbol: Vps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843818.3 | XP_004843875.1 | 3 - 4 | 282 | 81 | |
| XM_004843819.3 | XP_004843876.1 | 2 - 3 | 372 | 81 | ||
More Information
Set Membership: |
|

Vps29
Genomic Map