Assay Details
| Gene Symbol: Rita1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843743.3 | XP_004843800.1 | 2 - 3 | 757 | 67 | |
| XM_004843744.3 | XP_004843801.1 | 3 - 4 | 892 | 67 | ||
More Information
Set Membership: |
|

Rita1
Genomic Map