Assay Details
| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834917.3 | XP_004834974.1 | 5 - 6 | 1490 | 65 | |
| XM_004834918.3 | XP_004834975.1 | 5 - 6 | 1487 | 65 | ||
| XM_004834920.3 | XP_004834977.1 | 5 - 6 | 1935 | 65 | ||
| XM_021256374.1 | XP_021112033.1 | 5 - 6 | 1493 | 65 | ||
| XM_021256381.1 | XP_021112040.1 | 5 - 6 | 1490 | 65 | ||
More Information
Set Membership: |
|

Klhl24
Genomic Map