Assay Details
| Gene Symbol: Tppp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845117.2 | XP_004845174.1 | 2 - 3 | 398 | 81 | |
| XM_021258566.1 | XP_021114225.1 | 2 - 3 | 406 | 81 | ||
| XM_021258567.1 | XP_021114226.1 | 2 - 3 | 398 | 81 | ||
More Information
Set Membership: |
|

Tppp
Genomic Map