Assay Details
| Gene Symbol: Traf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852025.2 | XP_004852082.1 | 4 - 5 | 859 | 87 | |
| XM_013074828.2 | XP_012930282.1 | 4 - 5 | 858 | 87 | ||
| XM_013074829.2 | XP_012930283.1 | 4 - 5 | 854 | 87 | ||
| XM_021264336.1 | XP_021119995.1 | 4 - 5 | 854 | 87 | ||
More Information
Set Membership: |
|

Traf6
Genomic Map