Assay Details
| Gene Symbol: Lpar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841361.3 | XP_004841418.1 | 2 - 3 | 886 | 102 | |
| XM_013071065.2 | XP_012926519.1 | 2 - 3 | 936 | 102 | ||
More Information
Set Membership: |
|

Lpar3
Genomic Map