Assay Details
| Gene Symbol: Slc25a44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871190.2 | XP_004871247.1 | 2 - 3 | 811 | 84 | |
| XM_004871192.2 | XP_004871249.1 | 3 - 4 | 992 | 84 | ||
| XM_013067976.2 | XP_012923430.1 | 3 - 4 | 992 | 84 | ||
| XM_021249535.1 | XP_021105194.1 | 3 - 4 | 992 | 84 | ||
| XM_021249536.1 | XP_021105195.1 | 3 - 4 | 992 | 84 | ||
More Information
Set Membership: |
|

Slc25a44
Genomic Map