Assay Details
| Gene Symbol: Mafk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840351.3 | XP_004840408.1 | 3 - 4 | 3774 | 93 | |
| XM_004840352.3 | XP_004840409.1 | 2 - 3 | 259 | 93 | ||
| XM_021254635.1 | XP_021110294.1 | 3 - 4 | 346 | 93 | ||
| XM_021254637.1 | XP_021110296.1 | 4 - 5 | 3867 | 93 | ||
More Information
Set Membership: |
|

Mafk
Genomic Map