Assay Details
| Gene Symbol: Mapk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835394.3 | XP_004835451.1 | 3 - 4 | 1494 | 65 | |
| XM_004835395.3 | XP_004835452.1 | 2 - 3 | 1381 | 65 | ||
| XM_004835396.2 | XP_004835453.1 | 2 - 3 | 1247 | 65 | ||
| XM_021261083.1 | XP_021116742.1 | 2 - 3 | 2094 | 65 | ||
| XM_021261087.1 | XP_021116746.1 | 2 - 3 | 1299 | 65 | ||
| XM_021261094.1 | XP_021116753.1 | 1 - 2 | 556 | 65 | ||
| XM_021261097.1 | XP_021116756.1 | 2 - 3 | 1248 | 65 | ||
| XM_021261100.1 | XP_021116759.1 | 2 - 3 | 1250 | 65 | ||
More Information
Set Membership: |
|

Mapk6
Genomic Map