Assay Details
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866815.3 | XP_004866872.2 | 4 - 5 | 374 | 67 | |
XM_004866816.3 | XP_004866873.3 | 4 - 5 | 462 | 67 | ||
XM_004866817.3 | XP_004866874.3 | 4 - 5 | 469 | 67 | ||
XM_013066389.2 | XP_012921843.2 | 4 - 5 | 468 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
