Assay Details
| Gene Symbol: Sun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013070427.2 | XP_012925881.2 | 2 - 3 | 620 | 93 | |
| XM_013070428.2 | XP_012925882.1 | 2 - 3 | 1244 | 93 | ||
| XM_021254248.1 | XP_021109907.1 | 2 - 3 | 626 | 93 | ||
| XM_021254249.1 | XP_021109908.1 | 2 - 3 | 421 | 93 | ||
| XM_021254250.1 | XP_021109909.1 | 2 - 3 | 161 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013070428.2, XM_021254249.1, XM_021254250.1 transcripts.
Set Membership: |
|

Sun3
Genomic Map