Assay Details
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848459.3 | XP_004848516.1 | 6 - 7 | 658 | 64 | |
| XM_021261365.1 | XP_021117024.1 | 2 - 3 | 159 | 64 | ||
More Information
Set Membership: |
|

Ctsb
Genomic Map