Assay Details
| Gene Symbol: Mpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875185.3 | XP_004875242.1 | 5 - 6 | 600 | 62 | |
| XM_004875186.2 | XP_004875243.1 | 5 - 6 | 601 | 62 | ||
| XM_004875189.2 | XP_004875246.1 | 5 - 6 | 603 | 62 | ||
More Information
Set Membership: |
|

Mpp1
Genomic Map