Assay Details
| Gene Symbol: LOC101696409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834183.2 | XP_004834240.1 | 12 - 13 | 1123 | 91 | |
| XM_013070470.2 | XP_012925924.1 | 12 - 13 | 1120 | 91 | ||
More Information
Set Membership: |
|

LOC101696409
Genomic Map