Assay Details
| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862685.3 | XP_004862742.1 | 4 - 5 | 696 | 64 | |
| XM_004862686.3 | XP_004862743.1 | 4 - 5 | 692 | 64 | ||
| XM_004862687.3 | XP_004862744.1 | 3 - 4 | 522 | 64 | ||
More Information
Set Membership: |
|

Tfpi2
Genomic Map