Assay Details
| Gene Symbol: Prok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854206.2 | XP_004854263.1 | 1 - 2 | 288 | 89 | |
| XM_004854207.2 | XP_004854264.1 | 1 - 2 | 288 | 89 | ||
More Information
Set Membership: |
|

Prok2
Genomic Map