Assay Details
| Gene Symbol: LOC101713583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868637.3 | XP_004868694.1 | 1 - 2 | 208 | 73 | |
| XM_021247562.1 | XP_021103221.1 | 1 - 2 | 219 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101713583| Gene Symbol: LOC101713583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868637.3 | XP_004868694.1 | 1 - 2 | 208 | 73 | |
| XM_021247562.1 | XP_021103221.1 | 1 - 2 | 219 | 73 | ||