Assay Details
| Gene Symbol: LOC101713583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868637.3 | XP_004868694.1 | 2 - 3 | 477 | 67 | |
| XM_021247562.1 | XP_021103221.1 | 2 - 3 | 488 | 67 | ||
More Information
Set Membership: |
|

LOC101713583
Genomic Map