Assay Details
Gene Symbol: Rhbdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851011.2 | XP_004851068.1 | 3 - 4 | 661 | 65 | |
XM_004851012.2 | XP_004851069.1 | 3 - 4 | 561 | 65 | ||
XM_013074347.2 | XP_012929801.1 | 2 - 3 | 662 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
