Assay Details
| Gene Symbol: Ufd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843247.2 | XP_004843304.1 | 7 - 8 | 743 | 79 | |
| XM_004843248.3 | XP_004843305.1 | 7 - 8 | 676 | 79 | ||
| XM_021257044.1 | XP_021112703.1 | 6 - 7 | 576 | 79 | ||

Ufd1
Genomic Map