Assay Details
| Gene Symbol: Ufd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843247.2 | XP_004843304.1 | 8 - 9 | 803 | 67 | |
| XM_004843248.3 | XP_004843305.1 | 8 - 9 | 736 | 67 | ||
| XM_021257044.1 | XP_021112703.1 | 7 - 8 | 636 | 67 | ||
More Information
Set Membership: |
|

Ufd1
Genomic Map