Assay Details
| Gene Symbol: Plpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848515.3 | XP_004848572.1 | 3 - 4 | 914 | 101 | |
| XM_004848516.3 | XP_004848573.1 | 3 - 4 | 923 | 101 | ||
| XM_021261419.1 | XP_021117078.1 | 3 - 4 | 512 | 101 | ||
More Information
Set Membership: |
|

Plpp1
Genomic Map