Assay Details
| Gene Symbol: Stk25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868489.3 | XP_004868546.1 | 3 - 4 | 413 | 64 | |
| XM_004868490.3 | XP_004868547.1 | 3 - 4 | 410 | 64 | ||
| XM_004868491.3 | XP_004868548.1 | 3 - 4 | 409 | 64 | ||
| XM_021247359.1 | XP_021103018.1 | 2 - 3 | 259 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021247359.1 transcripts.
Set Membership: |
|

Stk25
Genomic Map