Assay Details
| Gene Symbol: Ap2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870653.3 | XP_004870710.1 | 16 - 17 | 2375 | 63 | |
| XM_004870654.3 | XP_004870711.1 | 16 - 17 | 2335 | 63 | ||
| XM_004870655.3 | XP_004870712.1 | 16 - 17 | 2288 | 63 | ||
| XM_004870656.3 | XP_004870713.1 | 15 - 16 | 2333 | 63 | ||
More Information
Set Membership: |
|

Ap2b1
Genomic Map